Skip to Content
Merck
All Photos(1)

Documents

SYNC195323C

Synthware chromatography column with spherical joints and coarse fritted disc

joint: ST/NS 35/20, I.D. × L 26.0 mm × 305 mm

Synonym(s):

Synthware C195323C, glass chromatograpy column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51

Joint

joint: ST/NS35/20

grade

for chromatography

packaging

pkg of 1 ea

manufacturer/tradename

Synthware C195323C

I.D. × L

26.0 mm × 305 mm

Looking for similar products? Visit Product Comparison Guide

General description

With a coarse porosity fritted disc to support packing material. All Synthware columns are heavy wall designed to increase safety.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service