Skip to Content
Merck
All Photos(1)

Key Documents

SIK41005-20

Sigma-Aldrich

1,4-Bis(trimethoxysilyl)benzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O6Si2
CAS Number:
Molecular Weight:
318.47
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

95%

form

liquid

shelf life

3 years under the recommended conditions

SMILES string

[Si](OC)(OC)(OC)c1ccc(cc1)[Si](OC)(OC)OC

InChI

1S/C12H22O6Si2/c1-13-19(14-2,15-3)11-7-9-12(10-8-11)20(16-4,17-5)18-6/h7-10H,1-6H3

InChI key

YIRZROVNUPFFNZ-UHFFFAOYSA-N

Application

This product is used to improve the dispersibility of modified silica powders into PDMS matrix by improving the adhesion between modified silica powders and the PDMS matrix.

Legal Information

Product of SiKEMIA

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service