Key Documents
CAD4112
Standard point hypodermic needles, 304 stainless steel
gauge 33, L 3/8 in., nickel plated hub
About This Item
UNSPSC Code:
42142523
NACRES:
NB.03
material
304 stainless steel needle
nickel plated hub
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4112
L
3/8 in.
gauge
33
Related Categories
General description
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Brad Nelms et al.
Science (New York, N.Y.), 364(6435), 52-56 (2019-04-06)
In multicellular organisms, the entry into meiosis is a complex process characterized by increasing meiotic specialization. Using single-cell RNA sequencing, we reconstructed the developmental program into maize male meiosis. A smooth continuum of expression stages before meiosis was followed by