BR780762
BRAND® micro tubes without tamper-evident screw cap
capacity 1.5 mL, conical bottom, non-sterile
Synonym(s):
Micro Tubes with Screw caps, Microcentrifuge Tubes, Tubes
All Photos(1)
About This Item
material
colorless tube
conical bottom
polypropylene tube
sterility
non-sterile
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 780762
capacity
1.5 mL
tube diam.
13 mm
General description
Operating range: -196 °C to +121 °C. Autoclavable at 121 °C (2 bar), acc. DIN EN 285.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG