Key Documents
BAF188900100
Scienceware® rack slanter
polypropylene, pack of 1 ea
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
Pricing and availability is not currently available.
material
polypropylene
feature
autoclavable
Holds 1 x rack
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18890-0100
size
293x242x83 mm
General description
Tilt your test tube rack to 25°, 35° or 45°. Adjusting the angle of racks reduces hand strain when working with pipettors. Increase the surface area of your Agar inside test tubes. Fits racks up 293 mm x 242 mm x 83 mm H (11 9/16 x 9 9/16 x 3 1/4 in. H).
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)