Skip to Content
Merck
All Photos(1)

Key Documents

B3159

Sigma-Aldrich

Potassium benzene-1,2-disulfonate

≥97%

Synonym(s):

Benzene-1,2-disulfonic acid dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(SO3K)2
CAS Number:
Molecular Weight:
314.42
Beilstein:
6552908
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

mp

≥300 °C (lit.)

SMILES string

[K+].[K+].[O-]S(=O)(=O)c1ccccc1S([O-])(=O)=O

InChI

1S/C6H6O6S2.2K/c7-13(8,9)5-3-1-2-4-6(5)14(10,11)12;;/h1-4H,(H,7,8,9)(H,10,11,12);;/q;2*+1/p-2

InChI key

WUERIJJOLNKVMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium benzene-1,2-disulfonate can be used to synthesize potassium 1-phenolate-4-disulfonate by reacting with potassium hydroxide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patai S
The Chemistry of sulphonic acids, esters, and their derivatives, 684-684 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service