AFM-71856-03
Muscovite Mica for AFM
(V-1 Quality), size 15 mm, pack of 10
Synonym(s):
Muscovite Mica for AFM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111761
NACRES:
NA.23
Recommended Products
form
solid
packaging
pack of 10
size
15 mm
thickness
0.15-0.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
Mica, 15mm diameter, thickness 0.15-0.2 mm, V-1 grade, Clear - Hard, of uniform color, nearly flat, free of all stains, foreign inclusion, cracks, and other similar defects. Perfect for AFM substrate
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service