A20804
5-Hydroxy-2-pentanone
Mixture of monomer and dimer, 95%
Synonym(s):
3-Acetyl-1-propanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COCH2CH2CH2OH
CAS Number:
Molecular Weight:
102.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.437 (lit.)
bp
144-145 °C/100 mmHg (lit.)
density
1.007 g/mL at 25 °C (lit.)
SMILES string
CC(=O)CCCO
InChI
1S/C5H10O2/c1-5(7)3-2-4-6/h6H,2-4H2,1H3
InChI key
JSHPTIGHEWEXRW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
199.4 °F - closed cup
Flash Point(C)
93 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
V V Neriueva et al.
Gigiena i sanitariia, (9)(9), 4-5 (1993-09-01)
Toxic properties of acetopropyl alcohol were studied. Symptoms of CNS damages predominated in toxicodynamics. A maximal single concentration is 0.2 mg/m3 (4th class of danger).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service