Skip to Content
Merck
All Photos(1)

Key Documents

926728

Sigma-Aldrich

PMPBO

≥95%

Synonym(s):

N-Benzyl-N-(5-methyl-[1,1′-biphenyl]-2-yl)oxalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N2O2
CAS Number:
Molecular Weight:
344.41
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

mp

154-155 °C

storage temp.

−20°C

SMILES string

O=C(C(NCC1=CC=CC=C1)=O)NC2=CC=C(C)C=C2C3=CC=CC=C3

InChI

1S/C22H20N2O2/c1-16-12-13-20(19(14-16)18-10-6-3-7-11-18)24-22(26)21(25)23-15-17-8-4-2-5-9-17/h2-14H,15H2,1H3,(H,23,25)(H,24,26)

InChI key

AWTLXLDETPWZRB-UHFFFAOYSA-N

Application

PMPBO is a ligand for the copper-catalyzed Ullman-Ma cross-coupling reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuntong Zhai et al.
The Journal of organic chemistry, 82(9), 4964-4969 (2017-04-22)
Diaryl formation is achieved by coupling phenols and (hetero)aryl halides under the catalysis of CuI/N,N'-bis(2-phenylphenyl) oxalamide (BPPO) or CuI/N-(2-phenylphenyl)-N'-benzyl oxalamide (PPBO) at 90 °C using DMF or MeCN as the solvent. Only 0.2-2 mol % CuI and ligand are required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service