Skip to Content
Merck
All Photos(1)

Key Documents

906573

Sigma-Aldrich

DLAM-Iδ1VproS-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

General description

DLAM-Iδ1VproS-13CH3 kit has 13CH3 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

DLAM-Iδ1VproS-13CH3 is an optimized kit to produce proteins with 13CH3 isotopomer on the methyl groups of isoleucine and valine. This kit has been tested with protein isotopic labeling in E. coli. It is used in NMR investigations of the structure, dynamics and function of challenging biological systems at atomic resolution. Introduction of 13C -labeled methyl group in highly deuterated background enhances the resolution of methyl-TROSY spectra.
For detection of long-range nOes or for the NMR study of large proteins

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heyi Zhang et al.
Current opinion in structural biology, 38, 75-82 (2016-06-14)
Proteins come together in macromolecular assemblies, recognizing and binding to each other through their structures, and operating on their substrates through their motions. Detailed characterization of these processes is particularly suited to NMR, a high-resolution technique sensitive to structure, dynamics
Rime Kerfah et al.
Current opinion in structural biology, 32, 113-122 (2015-04-17)
Nuclear magnetic resonance (NMR) spectroscopy is a uniquely powerful tool for studying the structure, dynamics and interactions of biomolecules at atomic resolution. In the past 15 years, the development of new isotopic labeling strategies has opened the possibility of exploiting
Stereospecific isotopic labeling of methyl groups for NMR spectroscopic studies of high-molecular-weight proteins.
Pierre Gans et al.
Angewandte Chemie (International ed. in English), 49(11), 1958-1962 (2010-02-17)
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service