Skip to Content
Merck
All Photos(2)

Key Documents

706418

Sigma-Aldrich

α-Cyclohexylmandelic acid

97%

Synonym(s):

2-Cyclohexyl-2-hydroxy-2-phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18O3
CAS Number:
Molecular Weight:
234.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

powder

mp

170-175 °C

functional group

carboxylic acid
hydroxyl
phenyl

SMILES string

OC(=O)C(O)(C1CCCCC1)c2ccccc2

InChI

1S/C14H18O3/c15-13(16)14(17,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12,17H,2,5-6,9-10H2,(H,15,16)

InChI key

YTRNSQPXEDGWMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Yoshida et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 111, 167-176 (2017-10-03)
Carboxylesterase (CES) and arylacetamide deacetylase (AADAC) are the major enzymes responsible for the hydrolysis of various clinical drugs. Our recent study demonstrated that the identity of the responsible hydrolase can be roughly surmised based on the chemical structures of compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service