Skip to Content
Merck
All Photos(1)

Key Documents

613924

Sigma-Aldrich

Chloroethane-2,2,2-d3

98 atom % D

Synonym(s):

Ethyl chloride-2,2,2-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CH2Cl
CAS Number:
Molecular Weight:
67.53
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCl

InChI

1S/C2H5Cl/c1-2-3/h2H2,1H3/i1D3

InChI key

HRYZWHHZPQKTII-FIBGUPNXSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 5 g quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 5 psig for 5 g. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - Repr. 1B

Storage Class Code

2A - Gases

WGK

WGK 2

Flash Point(F)

-58.0 °F - closed cup

Flash Point(C)

-50.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service