542822
4-(Trifluoromethyl)benzamidoxime
97%
Synonym(s):
N-Hydroxy-4-(trifluoromethyl)benzamidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CF3C6H4C(=NOH)NH2
CAS Number:
Molecular Weight:
204.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
125-130 °C (lit.)
functional group
amine
fluoro
oxime
SMILES string
N\C(=N/O)c1ccc(cc1)C(F)(F)F
InChI
1S/C8H7F3N2O/c9-8(10,11)6-3-1-5(2-4-6)7(12)13-14/h1-4,14H,(H2,12,13)
InChI key
QCVFLUSIBKAKPC-UHFFFAOYSA-N
Related Categories
Application
4-(Trifluoromethyl)benzamidoxime (N-Hydroxy-4-(trifluoromethyl)benzamidine) may be used to synthesize 5-(3-chlorothiophen-2-yl)-3-(4-trifluoromethylphenyl)-1,2,4-oxadiazole, a potential anticancer agent.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Han-Zhong Zhang et al.
Journal of medicinal chemistry, 48(16), 5215-5223 (2005-08-05)
We have identified 5-(3-chlorothiophen-2-yl)-3-(4-trifluoromethylphenyl)-1,2,4-oxadiazole (1d) as a novel apoptosis inducer through our caspase- and cell-based high-throughput screening assay. Compound 1d has good activity against several breast and colorectal cancer cell lines but is inactive against several other cancer cell lines.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service