All Photos(2)
About This Item
Linear Formula:
[CH3(CH2)17]2NOH
CAS Number:
Molecular Weight:
537.99
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
concentration
65%
mp
96-99 °C (lit.)
solubility
H2O: insoluble
SMILES string
CCCCCCCCCCCCCCCCCCN(O)CCCCCCCCCCCCCCCCCC
InChI
1S/C36H75NO/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37(38)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h38H,3-36H2,1-2H3
InChI key
ITUWQZXQRZLLCR-UHFFFAOYSA-N
Related Categories
Application
UV light stabilizer.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service