Skip to Content
Merck
All Photos(1)

Key Documents

364428

Sigma-Aldrich

(S)-(−)-Citronellic acid

98%

Synonym(s):

(S)-(−)-3,7-Dimethyl-6-octenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CO2H
CAS Number:
Molecular Weight:
170.25
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

optical activity

[α]25/D −8°, neat

refractive index

n20/D 1.453 (lit.)

bp

115-120 °C/0.4 mmHg (lit.)

density

0.926 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

C[C@@H](CC\C=C(/C)C)CC(O)=O

InChI

1S/C10H18O2/c1-8(2)5-4-6-9(3)7-10(11)12/h5,9H,4,6-7H2,1-3H3,(H,11,12)/t9-/m0/s1

InChI key

GJWSUKYXUMVMGX-VIFPVBQESA-N

Application

(S)-(-)-Citronellic acid may be used in the synthesis of trisubstituted piperidine substructures of the veratramine and jervine type, that can be building blocks for developing cyclopamine analogs.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Philipp Heretsch et al.
Organic letters, 11(23), 5410-5412 (2009-11-04)
All four diastereomers of the trisubstituted piperidine-alkaloids of the veratramine and jervine type were synthesized with complete stereocontrol starting from enantiopure citronellic acids. The flexible, high-yielding, and scalable route described here will facilitate convergent syntheses and give access to analogues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service