Skip to Content
Merck
All Photos(1)

Key Documents

17763

Sigma-Aldrich

(+)-Limonene 1,2-epoxide

mixture of cis/trans-isomers, ≥97.0% (sum of isomers, GC)

Synonym(s):

(+)-Limonene epoxide, (+)-p-Mentha-1,8-diene 1,2-epoxide, (4R)-1,2-Epoxy-p-menth-8-ene, (4R)-4-Isopropenyl-1-methyl-1-cyclohexene 1,2-epoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
80231
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (sum of isomers, GC)

form

liquid

refractive index

n20/D 1.467

density

0.930 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)[C@@H]1CCC2(C)OC2C1

InChI

1S/C10H16O/c1-7(2)8-4-5-10(3)9(6-8)11-10/h8-9H,1,4-6H2,2-3H3/t8-,9?,10?/m1/s1

InChI key

CCEFMUBVSUDRLG-XNWIYYODSA-N

General description

(+)-Limonene 1,2-epoxide, a monoterpene that can be prepared from (+)-limonene. It shows potent antinociceptive and antitumoral activity and is mainly used as a building block in synthesis of pharmaceutical products, perfumes, cosmetics and food additives.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Amanda C de Almeida et al.
Brain research, 1448, 56-62 (2012-03-01)
The aim of the study is to determine the acute toxicity and anxiolytic-like effects of a mixture of cis and trans of (+)-limonene epoxide in animal models of anxiety. After acute treatment with (+)-limonene epoxide at doses of 25, 50

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service