Skip to Content
Merck
All Photos(3)

Key Documents

114081

Sigma-Aldrich

4-(Benzyloxy)benzyl chloride

97%

Synonym(s):

4-Chloromethyl-α-phenylanisole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4CH2Cl
CAS Number:
Molecular Weight:
232.71
Beilstein:
882526
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

77-79 °C (lit.)

functional group

chloro
phenyl

storage temp.

2-8°C

SMILES string

ClCc1ccc(OCc2ccccc2)cc1

InChI

1S/C14H13ClO/c15-10-12-6-8-14(9-7-12)16-11-13-4-2-1-3-5-13/h1-9H,10-11H2

InChI key

UYQPSKUPEXAQRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Benzyloxy)benzyl chloride acts as a solid support to readily determine the loading of the corresponding polymer-bound acids by direct cleavage. This was determined while studying the coupling of substituted aromatic, heterocyclic, and alkyl carboxylates to the resin[1].

Application

4-(benzyloxy)benzyl chloride is used in the preparation of 4-aryl-3,4-dihydropyrimidine-5-carboxylates (DHPMs) which exhibits a wide spectrum of biological effects[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Speed Couplings and Cleavages in Microwave-Heated, Solid-Phase Reactions at High Temperatures.
The Journal of Organic Chemistry, 2001(5), 919-925 (2001)
C O Kappe
Bioorganic & medicinal chemistry letters, 10(1), 49-51 (2000-01-15)
A series of pharmacologically active, functionalized 4-aryl-3,4-dihydropyrimidine-5-carboxylates (DHPMs) are prepared by a versatile novel solid phase approach. In the key step, a polymer-bound thiouronium salt is condensed with unsaturated 1-ketoesters. The resulting polymer bound 1,4-dihydropyrimidines are cleaved from the resin
Jianbo Li et al.
Drug delivery, 25(1), 1117-1126 (2018-05-22)
Asthma is one of the most prevalent chronic inflammatory diseases of lung. Current asthma therapy using inhaled corticosteroid often results in undesired treatment outcome due to poor compliance and drugs' lack of tissue specificity. N,N,N'-trimethyl-N'-(2-hydroxyl-3-methyl-5-123Iiodobenzyl)-1,3-propanediamine (HIPD), a phenolic propanediamine derivative
Crist N Filer et al.
Journal of labelled compounds & radiopharmaceuticals, 62(1), 24-27 (2018-07-15)
The 2-step synthesis of [1-14 C]tyramine hydrochloride is described with the product being characterized by TLC, HPLC, and UV spectroscopy. Several methods are provided to purify [1-14 C]tyramine hydrochloride, and its storage and stability are also discussed.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service