Merck
Phytochemical study of Juglans regia L. leaves.
Phytochemistry (2017-09-11)
Sebastian Schwindl, Birgit Kraus, Jörg Heilmann
RESUMEN
A methanolic extract of Juglans regia L. leaves was fractioned by various chromatographic techniques yielding a total of 40 metabolites belonging to megastigmane, tetralone, phenylpropanoid, neolignane and juglone glycosides. Ten unknown megastigmane glucoside derivatives (juglanionosides A-K, 1-10) and six unknown tetralone glucoside derivatives (juglanosides J-O, 11-16) together with 24 known compounds - among them 16 described for the first time in Juglans - were isolated. As characteristic structural feature, the previously undescribed compounds showed acylation of the sugar units with sinapic, ferulic, coumaric, benzoic or salicylic acid. Their chemical structures were elucidated on the basis of 1D and 2D NMR techniques, HRESIMS as well as CD spectroscopy. Absolute stereochemistry was revealed by mild alkaline hydrolysis and comparison of CD and polarimetric data to literature values.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
(S)-(+)-2-Methylbutyric anhydride, 95%