Merck
Indoleamine 2,3 Dioxygenase (IDO) Expression and Activity in Relapsing-Remitting Multiple Sclerosis.
PloS one (2015-06-26)
Roberta Mancuso, Ambra Hernis, Simone Agostini, Marco Rovaris, Domenico Caputo, Dietmar Fuchs, Mario Clerici
RESUMEN
Interferon gamma (IFN-γ) production induces the transcription of indoleamine 2,3 dioxygenase (IDO) resulting in the reduction of T-cell activation and proliferation through the depletion of tryptophan and the elicitation of Treg lymphocytes. IDO was shown to be involved in the pathogenesis of autoimmune diseases; we investigated whether changes in IDO gene expression and activity could be indicative of onset of relapse in multiple sclerosis (MS) patients. IDO and interferon-γ (IFN-γ) gene expression, serum IDO activity (Kynurenine/Tryptophan ratio) and serum neopterin concentration--a protein released by macrophages upon IFN-γ stimulation--were measured in 51 individuals: 36 relapsing remitting (RR)-MS patients (21 in acute phase--AMS, 15 in stable phase--SMS) and 15 healthy controls (HC). PBMCs samples in AMS patients were collected before (BT-AMS) and during glucocorticoids-based therapy (DT-AMS). IDO expression was increased and IFN-γ was decreased (p<0.001) in BT-AMS compared to SMS patients. Glucocorticoids-induced disease remission resulted in a significant reduction of IDO and IFN-γ gene expression, IDO catalytic activity (p<0.001). Serum neopterin concentration followed the same trend as IDO expression and activity. Measurement of IDO gene expression and activity in blood could be a useful marker to monitor the clinical course of RR-MS. Therapeutic interventions modulating IDO activity may be beneficial in MS.