Merck
Isolation and Characterization of a Novel Imidacloprid-Degrading Mycobacterium sp. Strain MK6 from an Egyptian Soil.
Journal of agricultural and food chemistry (2015-05-02)
Mahrous M Kandil, Carmen Trigo, William C Koskinen, Michael J Sadowsky
RESUMEN
Thus far, only a small number and types of bacteria with limited ability in degrading imidacloprid have been reported. Also, genes regulating imidacloprid (IMDA) degradation have yet to be discovered. To study this in more detail, an enrichment technique was used to isolate consortia and pure cultures of IMDA-degrading bacteria. Through this approach, we successfully isolated a novel bacterium capable of completely degrading IMDA as a sole nitrogen source. The bacterium was subsequently identified as Mycobacterium sp. strain MK6 by sequence analysis of its 16S rRNA gene (Genbank accession number KR052814 ). BLASTn searches indicated that 16S rRNA gene from Mycobacterium sp. strain MK6 was 99% identical to several Mycobacterium spp. Mycobacterium sp. strain MK6 transformed 99.7% added IMDA (150 μg mL(-1)) in <2 weeks (t1/2 = 1.6 days) to 6-chloronicotinic acid (6-CNA) as its major metabolite. Although the isolated strain and mixed bacterial consortia were able to degrade IMDA, they failed to grow further on 6-CNA, indicating a lack of IMDA mineralization to carbon dioxide. Small amounts of the desnitro-olefin and desnitro-degradates of IMDA were observed during the incubation but did not accumulate in culture medium.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
6-Chloropyridine-3-carboxylic acid, 99%