Merck
  • Indoxyl sulfate signals for rapid mRNA stabilization of Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2 (CITED2) and suppresses the expression of hypoxia-inducible genes in experimental CKD and uremia.
Indoxyl sulfate signals for rapid mRNA stabilization of Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2 (CITED2) and suppresses the expression of hypoxia-inducible genes in experimental CKD and uremia.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2013-06-25)
Tetsuhiro Tanaka, Junna Yamaguchi, Yoshiki Higashijima, Masaomi Nangaku
RESUMEN
Chronic hypoxia in the tubulointerstitium serves as a final common pathway in progressive renal disease. Circumstantial evidence suggests that hypoxia-inducible factor (HIF)-1 in the ischemic tubules may be functionally inhibited in a chronic kidney disease (CKD) milieu. In this study, we hypothesized that indoxyl sulfate (IS), a uremic toxin, impairs the cellular hypoxic response. In human kidney (HK-2) proximal tubular cells, IS reduced the hypoxic induction of HIF-1 target genes. This effect was not associated with quantitative changes in the HIF-1α protein, but with functional impairment of the HIF-1α C-terminal transactivation domain (CTAD). Among factors that impeded the recruitment of transcriptional coactivators to the HIF-1αCTAD, IS markedly up-regulated Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2 (CITED2) through a mechanism of post-transcriptional mRNA stabilization involving the extracellular signal-regulated kinase (ERK) 1/2 pathway. In vivo, disproportionate expression of HIF target genes was demonstrated in several CKD models, which was offset by an oral adsorbent, AST-120. Furthermore, administration of indole reduced the induction of angiogenic, hypoxia-inducible genes in rats with experimental heart failure. Results of these studies reveal a novel role of IS in modulating the transcriptional response of HIF-1 and provide insight into molecular mechanisms underlying progressive nephropathies as well as cardiovascular complications.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Indoxyl sulfate potassium salt