Merck
Preference of beta-lactam formation in Cu(I)-catalyzed intramolecular coupling of amides with vinyl bromides.
Organic letters (2008-08-30)
Qiwu Zhao, Chaozhong Li
RESUMEN
A general and highly efficient synthesis of 4-alkylidene-2-azetidinones was achieved by the Cu(I)-catalyzed intramolecular C-N coupling of amides with vinyl bromides. This 4-exo ring closure was found to be fundamentally preferred over other modes (5-exo, 6-exo, and 6-endo) of cyclization under copper catalysis. Tandem C-N bond formation was then successfully developed to allow the convenient generation of medium-sized lactams.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vinyl bromide, 98%
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF