Merck
  • Rapid quantification of iodopropynyl butylcarbamate as the preservative in cosmetic formulations using high-performance liquid chromatography-electrospray mass spectrometry.
Rapid quantification of iodopropynyl butylcarbamate as the preservative in cosmetic formulations using high-performance liquid chromatography-electrospray mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2001-05-30)
M Frauen, H Steinhart, C Rapp, U Hintze
RESUMEN
A simple, rapid and reproducible method for identification and quantification of iodopropynyl butylcarbamate (IPBC) in different cosmetic formulations is presented. The determination was carried out using a high-performance liquid chromatography (HPLC) procedure on a reversed phase column coupled to a single quadrupole mass spectrometer (MS) via an electrospray ionization (ESI) interface. Detection was performed in the positive selected ion-monitoring mode. In methanol/water extracts from different cosmetic formulations a detection limit between 50 and 100 ng/g could be achieved. A routine analytical procedure could be set up with good quantification reliability (relative standard deviation between 0.9 and 2.9%).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%