Merck
Novel model to study the physiological effects of temporary or prolonged sex steroid deficiency in male mice.
American journal of physiology. Endocrinology and metabolism (2020-12-15)
Na Ri Kim, Rougin Khalil, Karel David, Leen Antonio, Dieter Schollaert, Ludo Deboel, Erik Van Herck, Nele Wardenier, Martine Cools, Brigitte Decallonne, Frank Claessens, Vanessa Dubois, Dirk Vanderschueren
RESUMEN
Sex steroids are critical for skeletal development and maturation during puberty as well as for skeletal maintenance during adult life. However, the exact time during puberty when sex steroids have the highest impact as well as the ability of bone to recover from transient sex steroid deficiency is unclear. Surgical castration is a common technique to study sex steroid effects in rodents, but it is irreversible, invasive, and associated with metabolic and behavioral alterations. Here, we used a low dose (LD) or a high dose (HD) of gonadotropin-releasing hormone antagonist to either temporarily or persistently suppress sex steroid action in male mice, respectively. The LD group, a model for delayed puberty, did not show changes in linear growth or body composition, but displayed reduced trabecular bone volume during puberty, which fully caught up at adult age. In contrast, the HD group, representing complete pubertal suppression, showed a phenotype reminiscent of that observed in surgically castrated rodents. Indeed, HD animals exhibited severely impaired cortical and trabecular bone acquisition, decreased body weight and lean mass, and increased fat mass. In conclusion, we developed a rodent model of chemical castration that can be used as an alternative to surgical castration. Moreover, the transient nature of the intervention enables to study the effects of delayed puberty and reversibility of sex steroid deficiency.NEW & NOTEWORTHY We developed a rodent model of chemical castration, which can be used as an alternative to surgical castration. Moreover, the transient nature of the intervention enables to study the effects of delayed puberty and reversibility of sex steroid deficiency.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Mouse IGFBP-3 ELISA Kit, for serum, plasma and cell culture supernatant