Merck
  • DMH-CBD, a cannabidiol analog with reduced cytotoxicity, inhibits TNF production by targeting NF-kB activity dependent on A2A receptor.
DMH-CBD, a cannabidiol analog with reduced cytotoxicity, inhibits TNF production by targeting NF-kB activity dependent on A2A receptor.
Toxicology and applied pharmacology (2019-02-24)
Rangel L Silva, Gabriela T Silveira, Carlos W Wanderlei, Nerry T Cecilio, Alexandre G M Maganin, Marcelo Franchin, Lucas M M Marques, Norberto P Lopes, José A Crippa, Francisco S Guimarães, José C F Alves-Filho, Fernando Q Cunha, Thiago M Cunha
RESUMEN
Cannabidiol (CBD) is a natural compound with psychoactive therapeutic properties well described. Conversely, the immunological effects of CBD are still poorly explored. In this study, the potential anti-inflammatory effects and underlying mechanisms of CBD and its analog Dimethyl-Heptyl-Cannabidiol (DMH-CBD) were investigated using RAW 264.7 macrophages. CBD and DMH-CBD suppressed LPS-induced TNF production and NF-kB activity in a concentration-dependent manner. Both compounds reduced the NF-kB activity in a μM concentration range: CBD (IC50 = 15 μM) and DMH-CBD (IC50 = 38 μM). However, the concentrations of CBD that mediated NF-kB inhibition were similar to those that cause cytotoxicity (LC50 = 58 μM). Differently, DMH-CBD inhibited the NF-kB activation without cytotoxic effects at the same concentrations, although it provokes cytotoxicity at long-term exposure. The inhibitory action of the DMH-CBD on NF-kB activity was not related to the reduction in IkBα degradation or either p65 (NF-kB) translocation to the nucleus, although it decreased p38 MAP kinase phosphorylation. Additionally, 8-(3-Chlorostyryl) caffeine (CSC), an A2A antagonist, reversed the effect of DMH-CBD on NF-kB activity in a concentration-dependent manner. Collectively, our results demonstrated that CBD reduces NF-kB activity at concentrations intimately associated with those that cause cell death, whereas DMH-CBD decreases NF-kB activity at non-toxic concentrations in an A2A receptor dependent-manner.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
8-(3-Chlorostyryl)caffeine, ≥98% (HPLC), solid