Merck
An Extensive Study of Coumarin Synthesis via Knoevenagel Condensation in Choline Chloride Based Deep Eutectic Solvents.
Current organic synthesis (2020-05-19)
Melita Lončarić, Martina Sušjenka, Maja Molnar
RESUMEN
In order to preserve the environment from harmful organic solvents, a synthesis of coumarin derivatives was performed in deep eutectic solvents, which are considered as "green" due to their characteristics. Choline chloride based deep eutectic solvents (DESs) were employed, both as solvents and as catalysts, in the synthesis of coumarin derivatives via Knoevenagel condensation. In order to find the best DES for coumarin synthesis, 20 DESs were tested for the reaction of salicylaldehyde and dimethyl malonate at 80 °C. Among the twenty tested deep eutectic solvents only five were adequate for this kind of synthesis. The best DES for this reaction was found to be the one composed of choline chloride:urea (1:2). Most coumarin compounds were obtained in good to excellent yield. Compounds 1g, 2g and 2p should be pointed out due to their yields of 85, 88 and 98 %, respectively. 3-Acetylcoumarins 5a, 5c, 5d, 5e, 5f and 5g were synthesized under ultrasound irradiation and were also obtained in excellent yields of 90, 95, 98, 93, 94 and 85 %, respectively. Series of coumarin derivatives were successfully synthesized, either in choline chloide:urea DES at 80 °C or in ultrasound-assisted reaction, from different salicylaldehydes and active methylene compounds. These "green" methods were found to be very effective in Knoevenagel condensation, while DES was recycled for several cycles without any significant influence on the product yield.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
3-Ethoxysalicylaldehyde, 97%