Merck
A Cell-Surface Membrane Protein Signature for Glioblastoma.
Cell systems (2017-04-04)
Dhimankrishna Ghosh, Cory C Funk, Juan Caballero, Nameeta Shah, Katherine Rouleau, John C Earls, Liliana Soroceanu, Greg Foltz, Charles S Cobbs, Nathan D Price, Leroy Hood
RESUMEN
We present a systems strategy that facilitated the development of a molecular signature for glioblastoma (GBM), composed of 33 cell-surface transmembrane proteins. This molecular signature, GBMSig, was developed through the integration of cell-surface proteomics and transcriptomics from patient tumors in the REMBRANDT (n = 228) and TCGA datasets (n = 547) and can separate GBM patients from control individuals with a Matthew's correlation coefficient value of 0.87 in a lock-down test. Functionally, 17/33 GBMSig proteins are associated with transforming growth factor β signaling pathways, including CD47, SLC16A1, HMOX1, and MRC2. Knockdown of these genes impaired GBM invasion, reflecting their role in disease-perturbed changes in GBM. ELISA assays for a subset of GBMSig (CD44, VCAM1, HMOX1, and BIGH3) on 84 plasma specimens from multiple clinical sites revealed a high degree of separation of GBM patients from healthy control individuals (area under the curve is 0.98 in receiver operating characteristic). In addition, a classifier based on these four proteins differentiated the blood of pre- and post-tumor resections, demonstrating potential clinical value as biomarkers.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Avanti
Lactosyl(β) Sphingosine (d18:1), Avanti Research - A Croda Brand 860542P, powder