Merck
  • Effect of Planarity of Aromatic Rings Appended to o-Carborane on Photophysical Properties: A Series of o-Carboranyl Compounds Based on 2-Phenylpyridine- and 2-(Benzo[b]thiophen-2-yl)pyridine.
Effect of Planarity of Aromatic Rings Appended to o-Carborane on Photophysical Properties: A Series of o-Carboranyl Compounds Based on 2-Phenylpyridine- and 2-(Benzo[b]thiophen-2-yl)pyridine.
Molecules (Basel, Switzerland) (2019-01-10)
Hyomin Jin, Seonah Kim, Hye Jin Bae, Ji Hye Lee, Hyonseok Hwang, Myung Hwan Park, Kang Mun Lee
RESUMEN
Herein, we investigated the effect of ring planarity by fully characterizing four pyridine-based o-carboranyl compounds. o-Carborane was introduced to the C4 position of the pyridine rings of 2-phenylpyridine and 2-(benzo[b]thiophen-2-yl)pyridine (CB1 and CB2, respectively), and the compounds were subsequently borylated to obtain the corresponding CN-chelated compounds CB1B and CB2B. Single-crystal X-ray diffraction analysis of the molecular structures of CB2 and CB2B confirmed that o-carborane is appended to the aryl moiety. In photoluminescence experiments, CB2, but not CB1, showed an intense emission, assignable to intramolecular charge transfer (ICT) transition between the aryl and o-carborane moieties, in both solution and film states. On the other hand, in both solution and film states, CB1B and CB2B demonstrated a strong emission, originating from π-π * transition in the aryl groups, that tailed off to 650 nm owing to the ICT transition. All intramolecular electronic transitions in these o-carboranyl compounds were verified by theoretical calculations. These results distinctly suggest that the planarity of the aryl groups have a decisive effect on the efficiency of the radiative decay due to the ICT transition.