Cambiar Vistas
| A ustedes/SKU | Disponibilidad | Precio |
|---|---|---|
250 ea | Comprobar disponibilidad del carrito | $30.200 |
Acerca de este artículo
NACRES:
NB.35
UNSPSC Code:
41121812
Material:
white polystyrene
Packaging:
pkg of 250 ea
Manufacturer/tradename:
Heathrow Scientific 120836
$30.200
Comprobar disponibilidad del carrito
Servicio técnico
¿Necesita ayuda? Nuestro equipo de científicos experimentados está aquí para ayudarle.
Permítanos ayudarlematerial
white polystyrene
packaging
pkg of 250 ea
manufacturer/tradename
Heathrow Scientific 120836
L × W × H
25 mm × 8.9 mm × 7.5 mm
General description
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material. They are made out of anti-static polystyrene (food grade compliant) and have a holding capacity of approximately 500 μL.
Features and Benefits
- Easy to Handle
- Large tabs make boats easy to use, position and hold, especially with forceps
- Superb Sample Recovery
- Inert material enables the boat and contents to be added to solutions to aid 100% sample recovery, where protocols allow
- Anti-static properties prevent sample retention from static
1 of 1
Este artículo | |||
|---|---|---|---|
| manufacturer/tradename Heathrow Scientific 120836 | manufacturer/tradename Heathrow Scientific HS1423C | manufacturer/tradename Heathrow Scientific HS1421C | manufacturer/tradename Heathrow Scientific HS1423CC |
| material white polystyrene | material square black polystyrene | material square blue polystyrene | material square black polystyrene |
| packaging pkg of 250 ea | packaging pk of 500 ea | packaging pk of 500 ea | packaging pk of 500 ea |
| L × W × H 25 mm × 8.9 mm × 7.5 mm | L × W × H (140 mm) × 140 mm × 22 mm | L × W × H (140 mm) × 140 mm × 22 mm | L × W × H (140 mm) × 140 mm × 22 mm |
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?