
β-Cyano-L-alanine

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing