Skip to Content
Merck

Skip To

1605588

USP

Rosavin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2E)-3-Phenylprop-2-en-1-yl 6-O-α-L-arabinopyranosyl-β-D-glucopyranoside, (2R,3R,4S,5S,6R)-2-[(E)-3-Phenylprop-2-enoxy]-6-([(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl)oxane-3,4,5-triol, (E)-3-Phenyl-2-propenyl 6-O-a-L-arabinopyranosyl-β-D-glucopyranoside, Rosavidin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
CLP 895,000

About This Item

Empirical Formula (Hill Notation):
C20H28O10
CAS Number:
Molecular Weight:
428.43
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CLP 895,000


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

rosavin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1CO[C@@H](OC[C@H]2O[C@@H](OC\C=C\c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H]1O

InChI

1S/C20H28O10/c21-12-9-28-19(17(25)14(12)22)29-10-13-15(23)16(24)18(26)20(30-13)27-8-4-7-11-5-2-1-3-6-11/h1-7,12-26H,8-10H2/b7-4+/t12-,13+,14-,15+,16-,17+,18+,19-,20+/m0/s1

InChI key

RINHYCZCUGCZAJ-IPXOVKFZSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rosavin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Rhodiola rosea Capsules
  • Powdered Rhodiola rosea Extract
  • Powdered Rhodiola rosea
  • Rhodiola rosea
  • Rhodiola crenulata Root and Rhizome Powder
  • Rhodiola crenulata Root and Rhizome Dry Extract

Biochem/physiol Actions

Rosavin is a well-known glycoside antioxidant from Rhadiola rosea (Golden Root, or Arctic Root) that exhibits antidepressant, anxiolytic and adaptogenic properties.
Rosavin is an antidepressant and anxiolytic; antiapoptotic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL89264SML0336PHL89903
Rosavin phyproof® Reference Substance

PHL89264

Rosavin

Rosarin phyproof® Reference Substance

PHL89903

Rosarin

manufacturer/tradename

USP

manufacturer/tradename

PhytoLab

manufacturer/tradename

-

manufacturer/tradename

PhytoLab

grade

pharmaceutical primary standard

grade

primary reference standard

grade

-

grade

primary reference standard

application(s)

pharmaceutical (small molecule)

application(s)

food and beverages

application(s)

-

application(s)

food and beverages

format

neat

format

neat

format

-

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

API family

rosavin

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Rosavin


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters