1570029
USP
Promethazine Related Compound B
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Isopromethazine hydrochloride, N,N,β-Trimethyl-10H-phenothiazine-10-ethanamine monohydrochloride, N,N-Dimethyl-2-(10H-phenothiazin-10-yl)propan-1-amine hydrochloride
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
promethazine
manufacturer/tradename
USP
application(s)
pharmaceutical
format
neat
SMILES string
S1c2c(cccc2)N(c3c1cccc3)C(CN(C)C)C.Cl
InChI
1S/C17H20N2S.ClH/c1-13(12-18(2)3)19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H
InChI key
RISCHQYUHLQSBU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Promethazine Hydrochloride
- Promethazine Hydrochloride Injection
- Promethazine Hydrochloride Oral Solution
- Promethazine Hydrochloride Tablets
Analysis Note
Other Notes
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service