Skip to Content
Merck
All Photos(1)

Key Documents

1466663

USP

N-Nitrosodiisopropylamine (NDIPA)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Isopropyl-N-nitrosoisopropylamine, DIPNA, NDIP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 1 mg (in 1 mL methanol)

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

single component solution

storage temp.

−20°C

SMILES string

N(N=O)(C(C)C)C(C)C

InChI

1S/C6H14N2O/c1-5(2)8(7-9)6(3)4/h5-6H,1-4H3

InChI key

AUIKJTGFPFLMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the USP Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.

Application

N-Nitrosodiisopropylamine (NDIPA) USP reference standard is intended for use only as specifically prescribed in the United States Pharmacopoeia.

It is also used to prepare standard, standard stock, nitrosamine RS stock, Nitrosamine standards stock solution mixture, and sensitivity stock solutions to determine NDIPA impurity in drug substances and drug products (valsartan, irbesartan, and losartan potassium etc) by chromatography according to general chapter 〈1469〉 of United States Pharmacopeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?1469? NITROSAMINE IMPURITIES
United States Pharmacopeia, 46(5) (2021)

Articles

An overview of nitrosamine impurity testing, worldwide regulations, and key considerations in filter selection for sample preparation and analysis.

This application note describes the LC-MS-based quantitative analysis of known nitrosamine impurities following procedures 1 and 3 as given in USP general chapter <1469>.

GC-MS method detects nitrosamines in Valsartan tablets, meeting US FDA guidelines for pharmaceutical quality control.

Related Content

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service