808148
TSKgel® ODS-80Tm (5 µm) HPLC Columns
L x I.D. 15 cm × 4.6 mm, C18 (octadecyl) phase
Synonym(s):
TSKgel® Reversed Phase HPLC Column
About This Item
Recommended Products
Product Name
TSKgel® ODS-80Tm HPLC Column, phase C18 (octadecyl), L × I.D. 15 cm × 4.6 mm, 5 μm particle size
material
stainless steel
Agency
suitable for USP L1
product line
TSKgel®
manufacturer/tradename
Tosoh ODS-80Tm, 08148
extent of labeling
15% Carbon, monolayer, endcapped
parameter
0-100% organic solvent
0-45 °C temperature
1.2 mL/min flow rate
210 bar max. pressure
technique(s)
HPLC: suitable
L × I.D.
15 cm × 4.6 mm
matrix
silica particle platform
matrix active group
C18 (octadecyl) phase
particle size
5 μm
pore size
100 Å pore diameter
operating pH
2.0-7.5
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
General description
Physical form
Legal Information
guard cartridge
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The major research interests of Prof. Bing-Feng Shi's lab are focused on transition metal-catalyzed transformations and their application in the synthesis of natural products.
Yu program focuses on efficient C–H bond activation for drug synthesis, using simple starting materials.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service