57032
Visiprep™ Manifold Flow Control Valve
for use with standard 12- and 24-port models, pk of 2
Synonym(s):
V.M.FLOW CONTROL VALVES 2/PK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
product line
Visiprep™
packaging
pk of 2
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with standard 12- and 24-port models
Looking for similar products? Visit Product Comparison Guide
General description
Flow control valve can help in manipulating flow through each tube by rotating individual screw-valves built into the cover.
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service