507482
Septa, PTFE/Silicone
red PTFE/white silicone, diam. 12 mm, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
material
PTFE face
red PTFE/white silicone
silicone body
packaging
pkg of 100 ea
manufacturer/tradename
Chromacol 12-ST2
parameter
250 °C temp. limit
O.D.
12 mm
diam.
12 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
It is used as an air-tight seal, and prevents contamintion
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service