Skip to Content
Merck
All Photos(8)

Key Documents

ZRB2345

Sigma-Aldrich

Anti-PAX7 Antibody, clone 2D12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HuP1, Paired box protein Pax-7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2D12, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... PAX7(5081)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2D12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects PAX7. It targets an epitope within 25 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the C-terminal half of human Paired box protein Pax-7 (PAX7).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in Human testis tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected PAX7 in Human testis tissue sections.

Tested Applications

Enzyme Immunoassay (ELISA) Analysis: A serial of dilutions from a representative lot detected GST/His-tagged recombinant Human PAX7.

Affinity Binding Assay: A representative lot of this antibody bound PAX7 peptide with a KD of 1.4 x 10-7 in an affinity binding assay.

Flow Cytometry Analysis: 1 µg from a representative lot detected PAX7 in one million HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Paired box protein Pax-7 (UniProt: P23759; also known as HuP1, PAX7) is encoded by the PAX7 (also known as HUP1) gene (Gene ID: 5081) in human. PAX7 is a transcription factor that is involved in the regulation of muscle stem cells proliferation and plays a role in myogenesis and muscle regeneration. It can bind to DNA as a heterodimer with PAX3. Its homeobox DNA-binding domain is localized in amino acids 217-276. PAX7 has two acetylation sites (Lys 105 and Lys 193) that positively regulate its transcriptional activity and lack of acetylation reduces its DNA binding. Its acetylation levels are regulated by MYST1 and SIRT2. PAX7 expression is observed in quiescent as well as activated satellite cells, and its levels are downregulated when satellite cells commit to muscle differentiation. It is also critical for the normal function of satellite cells in adult skeletal muscle. PAX7 deficiency results in cell-cycle arrest and early differentiation.  A chromosomal aberration involving PAX7 is found in cases of rhabdomyosarcoma, a highly malignant tumor of striated muscle derived from primitive mesenchymal cells. Three isoforms of PAX7 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Sincennes, M-C. et al. (2021).  Nat. commun. 12; Article 3253; von Maltzahn, J., et al. (2013). Proc. Natl. Acad. Sci. USA. 110(41); 16474-16479; Sambasivan, R., et al.  (2011). Development. 138 (17): 3647-3656).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service