Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1597

Sigma-Aldrich

Anti-FOXO3A Antibody, clone 1G18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Forkhead box protein O3;AF6q21 protein;Forkhead in rhabdomyosarcoma-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G18, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 71.28 and 48.41 kDa
observed mol wt ~100 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G18 is a Rabbit recombinant monoclonal antibody that specifically detects Forkhead box protein O3 (FOXO3A). It targets an epitope within 20 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal half of human Forkhead box protein O3 (FOXO3A).

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FOXO3A in MCF-7 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected FOXO3A in SH-SY5Y cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected FOXO3A in human colon tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot from a representative lot detected FOXO3A in SH-SY5Y cells.

Affinity Binding Assay: A representative lot of this antibody bound FOXO3A with a KD of 3.0 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Forkhead box protein O3 (UniProt: O43524; also known as AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1) is encoded by the FOXO3 (also known as FKHRL1, FOXO3A) gene (Gene ID: 2309) in human. Forkhead box protein O3 is a transcriptional activator that triggers apoptosis in the absence of survival factors, including neuronal cell death upon oxidative stress. It recognizes and binds to the DNA sequence 5′-[AG]TAAA[TC]A-3′. Its DNA binding domain located in amino acids 157-251. The nuclear localization signal is localized in amino acids 242-259. In the cytoplasm it is present in an inactive state and it translocates to the nucleus upon oxidative stress and in the absence of survival factors. In response to metabolic stress, it translocates into the mitochondria where it promotes mtDNA transcription. In the presence of survival factors such as IGF-1, it is phosphorylated on threonine 32 and serine 253 by AKT1/PKB. The phosphorylated form then interacts with 14-3-3 proteins and is retained in the cytoplasm. Survival factor withdrawal induces dephosphorylation and promotes its translocation to the nucleus where the dephosphorylated protein induces transcription of target genes and triggers apoptosis. FOXO3 deacetylation by SIRT1 or SIRT2 stimulates its interaction SKP2 and facilitates SCF(SKP2)-mediated FOXO3 ubiquitination and proteasomal degradation. A chromosomal aberration involving FOXO3 is found in secondary acute leukemias. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Peserica, A., et al. (2013). Cell Mol. Life Sci. 70(11); 2015-2029; Brunet, A., et al. (1999). Cell. 96(6); 857-868).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service