Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1433

Sigma-Aldrich

Anti-SNAI1 Antibody, clone 1F16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
CLP 207,000
4 X 25 μL
CLP 428,000

CLP 207,000


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
CLP 207,000
4 X 25 μL
CLP 428,000

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CLP 207,000


Estimated to ship onMay 21, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F16, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 29.08 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

monkey, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F16 is a Rabbit recombinant monoclonal antibody that specifically detects Zinc finger protein SNAI1. It targets an epitope within 19 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the N-terminal region of human Zinc finger protein SNAI1.

Application

Quality Control Testing

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected SNAI1 in HEK293 cell lysate.

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected SNAI1 in lysates from Human kidney, NIH3T3, and Cos-7 cells..

Immunocytochemistry Analysis: 1:100 from a representative lot detected SNAI1 in NIH3T3 cells.

Affinity Binding Assay: A representative lot of this antibody bound SNAI1 with a KD of 2.9 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SNAI1, clone 1F16 ZooMAb®, Cat. No. ZRB1433, is a recombinant Rabbit monoclonal antibody that detects Zinc finger protein SNAI1 and is tested for use in, Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

Zinc finger protein SNAI1 (UniProt: O95863; also known as Protein snail homolog 1, Protein sna, SNAI1) is encoded by the SNAI1 (also known as SNAH) gene (Gene ID: 6615) in human. SNAI1 is expressed in a variety of tissue with highest expression observed in kidneys. It is also expressed in mesenchymal and epithelial cell lines. It contains four C2H2 type zinc-finger domains. SNAI1 is involved in induction of the epithelial to mesenchymal transition (EMT), formation and maintenance of embryonic mesoderm, growth arrest, survival, and cell migration. It has a SNAG domain (aa 1-20) that competes with histone H3 for the same binding site on the histone demethylase complex formed by KDM1A and RCOR1, and thereby inhibits demethylation of histone H3 at lysine 4. During EMT, it is involved with lysyl oxidase like 2 (LOXL2) in negatively regulating pericentromeric heterochromatin transcription. It recruits LOXL2 to pericentromeric regions to oxidize histone H3 and repress transcription, which leads to release of heterochromatin component CBX5/HP1A that enables chromatin reorganization and acquisition of mesenchymal traits. SNAI1 is localized both in the cytoplasm and nucleus. Upon phosphorylation by GSK-3b on multiple serine residues in the N-terminal region, it is it is exported from the nucleus to the cytoplasm where subsequent phosphorylation of the destruction motif and ubiquitination by b-transducin repeat containing E3 ubiquitin protein ligase (BTRC) occurs. Phosphorylation by casein kinase 1e, probably at serine 104, provides the priming site for the subsequent phosphorylation by GSK-3b. Its phosphorylation by p21 activated kinase (PAK1) may modulate its transcriptional activity by promoting increased accumulation in the nucleus. Phosphorylation at serine 11 and serine 92 are reported to positively regulate its functions in the induction of EMT and cell survival, respectively. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service