Skip to Content
Merck
All Photos(1)

Key Documents

Z370835

Sigma-Aldrich

Cuvette mixer

Narrow footprint

Synonym(s):

cuvettes, micro cuvette, plastic cuvette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
SB.32

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

Looking for similar products? Visit Product Comparison Guide

General description

for semi-micro cells

Features and Benefits

Up and down plunging motion provides thorough mixing in spectrophotometer cells. Clear acrylic with straight handle; two different "footprints" for full-size and semi-micro cells with 10 mm light path.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service