Z332380
Bottle adapters for BRAND® Dispensers and Burettes
GL 45 thread, GL 32 bottle, ETFE
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
Recommended Products
material
ETFE
manufacturer/tradename
BRAND 704398
fitting
GL 32 bottle
GL 45 thread
Related Categories
General description
For all Dispensette® models as well as seripettor® and QuikSip™
Other Notes
BR704398 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)