T1878
Trizma® Pre-Set crystals
Type 8.3-FT, pH 8.3, suitable for use with marine species
Synonym(s):
Tris crystals
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352104
NACRES:
NA.25
Recommended Products
type
Type 8.3-FT
Quality Level
form
crystalline powder
pH
8.3
solubility
water: 0.66 g/mL, clear, colorless
suitability
suitable for use with marine species
Application
Trizma® Pre-Set crystals is suitable for use with marine species.
Legal Information
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service