Skip to Content
Merck
All Photos(1)

Key Documents

SRP3072

Sigma-Aldrich

IL-11 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

AGIF (Adipogenesis inhibitory factor)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CLP 358,000

CLP 358,000


Estimated to ship onApril 27, 2025



Select a Size

Change View
10 μG
CLP 358,000

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

CLP 358,000


Estimated to ship onApril 27, 2025


biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤2.0 ng/mL ED50

mol wt

19.1 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL11(3589)

General description

IL-11 is a multifunctional cytokine produced by stromal cells such as fibroblasts, epithelial cells and osteoclasts. It is expressed in a wide variety of tissues including thymus, lung, bone, connective tissue and central nervous system. IL-11 plays an important regulatory role in hematopoiesis by stimulating growth of myeloid, erythroid and megakaryocyte progenitor cells. It also regulates bone metabolism, inhibits production of proinflammatory cytokines and protects against gastromucosal injury. Recombinant Human IL-11 is a 19.1 kDa protein consisting of 179 amino acid residues.

Biochem/physiol Actions

IL-11 is a multifunctional cytokine produced by stromal cells such as fibroblasts, epithelial cells and osteoclasts. Recombinant Human IL-11 is a 19.1 kDa protein consisting of 179 amino acid residues.

Sequence

MPGPPPGPPR VSPDPRAELD STVLLTRSLL ADTRQLAAQL RDKFPADGDH NLDSLPTLAM SAGALGALQL PGVLTRLRAD LLSYLRHVQW LRRAGGSSLK TLEPELGTLQ ARLDRLLRRL QLLMSRLALP QPPPDPPAPP LAPPSSAWGG IRAAHAILGG LHLTLDWAVR GLLLLKTRL

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Please Note: This solution is slow to dissolve. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fitzgerald, K.A., et al.
The Cytokine Factsbook, 95-98 (2001)
V F Quesniaux et al.
International review of experimental pathology, 34 Pt A, 205-214 (1993-01-01)
The novel stromal cell factor, IL-11, has been reported to have diverse effects on the lymphopoietic and myeloid/erythroid cells in vitro. These include expansion of T cell-dependent Ig-secreting B cells, proliferation and differentiation of megakaryocytic progenitors and of a variety
You-Jee Jang et al.
Reproduction, fertility, and development, 29(12), 2437-2445 (2017-05-20)
The aim of the present study was to examine the regulation of interleukin (IL)-11 expression, as well as the role of IL-11, during ovulation in gonadotropin-primed immature rats. Injection of equine chorionic gonadotropin (eCG), followed by human CG (hCG) to
Ji-Hong Lim
The Korean journal of physiology & pharmacology : official journal of the Korean Physiological Society and the Korean Society of Pharmacology, 18(5), 391-396 (2014-10-30)
Although interleukin-11 (IL-11) has been reported to be elevated in hypoxic tumors and has been associated with a poor prognosis in various cancers, little is known about its precise role in promoting metastasis in hypoxic tumors. In the present study
Javier Milara et al.
Respiratory research, 23(1), 313-313 (2022-11-16)
Pulmonary hypertension (PH) associated to idiopathic pulmonary fibrosis (IPF) portends a poor prognosis. IL-11 has been implicated in fibrotic diseases, but their role on pulmonary vessels is unknown. Here we analyzed the contribution of IL-11 to PH in patients with

Articles

Role of growth factors in stem cell differentiation and various growth factors for your research at sigmaaldrich.com

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service