Skip to Content
Merck
All Photos(1)

Key Documents

SML2493

Sigma-Aldrich

VX-702

≥98% (HPLC)

Synonym(s):

6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12F4N4O2
CAS Number:
Molecular Weight:
404.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1c(c(ccc1)F)N(c2nc(c(cc2)C(=O)N)c3c(cc(cc3)F)F)C(=O)N

InChI

1S/C19H12F4N4O2/c20-9-4-5-10(14(23)8-9)16-11(18(24)28)6-7-15(26-16)27(19(25)29)17-12(21)2-1-3-13(17)22/h1-8H,(H2,24,28)(H2,25,29)

InChI key

FYSRKRZDBHOFAY-UHFFFAOYSA-N

Biochem/physiol Actions

Second generation p38α MAPK inhibitor
VX-702 is a second generation p38α MAPK inhibitor with 14-fold higher potency against p38α versus p38β.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Athan Kuliopulos et al.
Thrombosis and haemostasis, 92(6), 1387-1393 (2004-12-08)
Systemic inflammation has been shown to be a contributing factor to the instability of atherosclerotic plaques in patients with acute coronary syndromes (ACS). VX-702, a novel p38 mitogen-activated protein kinase (MAPK) inhibitor, is currently under investigation in ACS patients with
Sen Han et al.
American journal of cancer research, 8(3), 489-501 (2018-04-11)
NK1.1-CD4+NKG2D+ cells exert their immune-regulatory function in tumor as an unconventional regulatory T cell subset through the production of TGF-β1; however, the molecular mechanisms involving with the activation of nuclear factors for TGF-β1 transcription remain unclear. Here we determined that
Ethan L Morgan et al.
PLoS pathogens, 14(4), e1006975-e1006975 (2018-04-10)
Human papillomaviruses (HPV) activate a number of host factors to control their differentiation-dependent life cycles. The transcription factor signal transducer and activator of transcription (STAT)-3 is important for cell cycle progression and cell survival in response to cytokines and growth
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service