Skip to Content
Merck
All Photos(1)

Key Documents

SML1195

Sigma-Aldrich

Palonosetron hydrochloride

≥98% (HPLC)

Synonym(s):

(3aS)-2-[(3S)-Quinuclidin-3-yl]-2,3,3a,4,5,6-hexahydro-1H-benzo[de]isoquinolin-1-one hydrochloride, (3aS)-2-[(3S)-1-Azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-1H-Benz[de]isoquinolin-1-one hydrochloride (1:1), RS 25259-197

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CLP 103,000
50 MG
CLP 406,000

CLP 103,000


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
CLP 103,000
50 MG
CLP 406,000

About This Item

Empirical Formula (Hill Notation):
C19H24N2O · HCl
CAS Number:
Molecular Weight:
332.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

CLP 103,000


Estimated to ship onMay 15, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -90 to -110°, c = 0.4 in H2O

storage condition

desiccated
protect from light

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1N([C@H]2C3CCN(CC3)C2)C[C@]4([H])C5=C1C=CC=C5CCC4.[H]Cl

InChI

1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m1./s1

InChI key

OLDRWYVIKMSFFB-SSPJITILSA-N

Gene Information

human ... HTR3A(3359)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Palonosetron is a potent and selective 5-HT3 receptor antagonist (Aloxi) used in the prevention and treatment of chemotherapy-induced nausea and vomiting (CINV).
Palonosetron is a potent and selective 5-HT3 receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Ma et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 13560-13571 (2019-10-02)
So far, a comprehensive animal model that can mimic both the central and peripheral pathophysiological changes of irritable bowel syndrome (IBS) is lacking. Here, we developed a novel IBS rat model combining trinitro-benzene-sulfonic acid (TNBS) and chronic unpredictable mild stress

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service