Skip to Content
Merck
All Photos(1)

Key Documents

SML0733

Sigma-Aldrich

Cilomilast

≥95% (HPLC)

Synonym(s):

SB 207499, SB-207499, cis-4-Cyano-4-[3-(cyclopentyloxy)-4-methoxyphenyl]cyclohexane-1-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25NO4
CAS Number:
Molecular Weight:
343.42
UNSPSC Code:
41106305
NACRES:
NA.77

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

−20°C

InChI

1S/C20H25NO4/c1-24-17-7-6-15(12-18(17)25-16-4-2-3-5-16)20(13-21)10-8-14(9-11-20)19(22)23/h6-7,12,14,16H,2-5,8-11H2,1H3,(H,22,23)

InChI key

CFBUZOUXXHZCFB-UHFFFAOYSA-N

Biochem/physiol Actions

Cilomilast (SB-207499) is a potent, selective and orally available PDE4 inhibitor that exhibits anti-inflamatory and anti-asthmatic activity.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simone C da Silva Rosa et al.
Autophagy, 17(9), 2257-2272 (2020-10-13)
Lipotoxicity is a form of cellular stress caused by the accumulation of lipids resulting in mitochondrial dysfunction and insulin resistance in muscle. Previously, we demonstrated that the mitophagy receptor BNIP3L/Nix is responsive to lipotoxicity and accumulates in response to a
Hongyan Ma et al.
Biochemical and biophysical research communications, 450(4), 1560-1567 (2014-07-16)
Acute lung injury (ALI), acute respiratory distress syndrome (ARDS), is actually involved in an ongoing and uncontrolled inflammatory response in lung tissues. Although extensive studies suggested that phospodiesterase type 4B (PDE4B) may be related to inflammation, the underlying cell biological

Articles

Cyclic nucleotide phosphodiesterases (PDEs) catalyze the hydrolysis of cAMP and/or cGMP. There are 11 different mammalian PDE families.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service