Recommended Products
biological source
plant
Assay
≥90% (LC/MS-ELSD)
form
solid
mol wt
388.45
solubility
water: slightly soluble
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
SMILES string
O1[C@@H]([C@H](C(=O)c2c1c(c(c(c2O)C(CCC)CC(=O)O)OC)CC=C(C)C)C)C
InChI key
IGEOTNLFIQJPLF-GRKKQISMSA-N
Related Categories
General description
Isocalolongic acid is a natural plant metabolite belonging to the class of pyranochromanone acids. It is commonly found in plants of Calophyllum species and is known for its potent antifungal and antimicrobial activity.
Application
It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.
Features and Benefits
- High quality compound suitable for multiple research applications
- Compatible with HPLC and mass spectrometry techniques
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Planta medica, 69(12), 1130-1135 (2004-01-30)
Two new xanthones, caledonixanthone M 1 and caloxanthone L 2, and one new acid, caledonic acid 6 were isolated from the hexane-soluble extract of the stem bark of Calophyllum caledonicum. In the course of this phytochemical study, seven other known
Phytotherapy research : PTR, 29(12), 1991-1994 (2015-10-31)
The Calophyllum genus is well-known for its antimicrobial and cytotoxic activities, and therefore, we analyzed these biological activities for resins of Calophyllum antillanum and Calophyllum inophyllum growing in Cuba. C. antillanum resins showed a potent activity against Plasmodium falciparum (IC50 = 0.3 ± 0.1 µg/mL)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service