Skip to Content
Merck
All Photos(6)

Key Documents

SAB5700677

Sigma-Aldrich

Anti-CTGF antibody produced in rabbit

Synonym(s):

CCN2, CTGF, HCS24, IGFBP8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CLP 349,000

CLP 349,000


Estimated to ship onApril 29, 2025



Select a Size

Change View
100 μL
CLP 349,000

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CLP 349,000


Estimated to ship onApril 29, 2025


biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35
38

species reactivity

mouse, rat, human

concentration

3.97 mg/ml

technique(s)

immunofluorescence: 1:20-1:50
immunohistochemistry: 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CTGF(1490)

General description

The protein encoded by this gene is a mitogen that is secreted by vascular endothelial cells. The encoded protein plays a role in chondrocyte proliferation and differentiation, cell adhesion in many cell types, and is related to platelet-derived growth factor. Certain polymorphisms in this gene have been linked with a higher incidence of systemic sclerosis.

Immunogen

Recombinant fusion protein containing a sequence corresponding to amino acids 27-349 of human CTGF (NP_001892.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service