Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500405

Sigma-Aldrich

Anti-Caspase-9 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Casp-9, Anti-Mch6/ Apaf-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 46 kDa

species reactivity

mouse, rat, human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP9(842)

Immunogen

Caspase-9 antibody was raised against a peptide corresponding to amino acids 41 to 56 of human caspase-9 .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500405.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Feng et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 52, 136-146 (2019-01-03)
Peeled stems of Syringa pinnatifolia Hemsl. (SP) have been widely used to treat extra "He-Yi" induced myocardial ischemia for hundreds of years in Inner Mongolia, China and previous result showed that intragastric pretreatment with total extract (T) of SP has
Xiaofen Zhang et al.
Biological chemistry (2019-03-22)
Pancreatitis is a disease caused by inflammation of pancreatic acinar cells. Geniposide (GEN) possesses anti-inflammation activities. Hence, we investigated the effects of GEN on lipopolysaccharide (LPS)-stimulated AR42J cells. AR42J cells were stimulated by LPS and then treated with GEN and/or
Fan Deng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(1), 279-292 (2017-11-14)
Hearts from diabetic subjects are susceptible to myocardial ischemia reperfusion (I/R) injury. Propofol has been shown to protect against myocardial I/R injury due to its antioxidant properties while the underlying mechanism remained incompletely understood. Thus, this study aimed to determine
Xiaofen Li et al.
EBioMedicine, 39, 159-172 (2018-12-12)
Ubiquitin-proteasome system (UPS) is integral to cell survival by maintaining protein homeostasis, and its dysfunction has been linked to cancer and several other human diseases. Through counteracting ubiquitination, deubiquitinases (DUBs) can either positively or negatively regulate UPS function, thereby representing
Mutation-Driven S100A8 Overexpression Confers Aberrant Phenotypes in Type 1 CALR-Mutated MPN.
Wang, et al.
International Journal of Molecular Sciences, 24 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service