Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Check Cart for Availability | CLP 660,000 |
About This Item
NACRES:
NA.32
UNSPSC Code:
41116158
Species reactivity:
bovine
Storage temp.:
−20°C
Shipped in:
wet ice
CLP 660,000
Check Cart for Availability
species reactivity
bovine
technique(s)
ELISA: suitable
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.12 ng/ml
shipped in
wet ice
storage temp.
−20°C
General description
This ELISA antibody pair detects Bovine IFN alpha (IFNA1)
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| species reactivity bovine | species reactivity bovine | species reactivity porcine | species reactivity bovine |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




